NOB ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:55894-52-5
:
Formula:C₁₅H₁₉NO₆
MDL Number:MFCD00132973
:309.318 g/mol
Storage Temperature:Ambient
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together