Order Entry
Sweden
ContactUsLinkComponent
Vacu-vials® Film Forming Amines Kit
Vacu-vials® Film Forming Amines Kit
Catalog # CHETK-1013
Supplier:  CHEMetrics
CAS Number:  
Vacu-vials® Film Forming Amines Kit
Catalog # CHETK-1013
Supplier:  CHEMetrics
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Vacu-vials® film forming amines (FFA) kit, 13 mm, Rose Bengal method
  • Range:
    0 - 6,00 ppm as ODA

Specifications

About this item

Film-forming amines, also known as FFAP or FFP, are chemicals applied in high-pressure boilers, cooling systems, and HRSGs to guard against corrosion and reduce the presence of corrosive agents.

  • Analysis time: 2 minutes

The test kit utilises the Rose Bengal method to detect and measure these substances in boiler water or steam condensate.

The kit comes in a cardboard box and contains everything needed to perform 30 tests: 30 ampoules, acidifier solution, 25 ml sample cup, ampoule blank, and instructions.

This kit requires the use of a V-3000 multi-analyte photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell.