Refractive Index Liquids, Series B
Catalog # CARG1812Y1.700
Supplier: CARGILLE
CAS Number:
Specifications
- Pk:30 mL
- Refractive index:1,700 nD
Specifications
About this item
Refractive index liquids have a broad range of applications in diversified fields.
- Light yellow brown liquid
- Slight unpleasant odour
Adjustment: ±0,0002
Choose from range nD 1,642 to 1,700