Barium perchlorate
Catalog # GFSC8
Supplier: GFS CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:336.23 g/mol
Merck Index:12,01016
Melting Pt:505 °C
Density:3.2 g/cm³ (20 °C)
Flash Pt:21 °C (closed cup)
EINECS:236-710-4
UN:1447
ADR:5.1,II
:GHS03
:GHS07
CAS Number:13465-95-7
Formula:Ba(ClO₄)₂
MDL Number:MFCD00003440