0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
L-α-Phenylalanine
L-Phenylalanine, Millipore®
L-Phenylalanine, Millipore®
:
Merck
L-Phenylalanine, Millipore®
:
Merck
:
63-91-2
:
:
C₉H₁₁NO₂
:
MFCD00064227
:
165.19 g/mol
:
14,07271
:
313 °C (1013 hPa)
:
173 °C
:
0.754 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
Ambient
:
200-568-1
msds_107256
ProductPage_107256