0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Silver nitrate
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF0611000
:
FF CHEMICALS
:
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF0611000
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
169.87 g/mol
:
Ambient
:
231-853-9
:
3082
:
9,III
:
GHS07
:
GHS09
:
7761-88-8
:
AgNO₃
:
MFCD00003414