Order Entry
Sweden
ContactUsLinkComponent
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Irritant
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF7115000
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Irritant
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF7115000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3082
ADR:9,III
 :GHS07
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414