Order Entry
Sweden
ContactUsLinkComponent
Silver nitrate 0.01 mol/l (0.01 N) in aqueous solution
Silver nitrate 0.01 mol/l (0.01 N) in aqueous solution
Catalog # FFCHFF4751000
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Silver nitrate 0.01 mol/l (0.01 N) in aqueous solution
Catalog # FFCHFF4751000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414