Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid 33%
Hydrochloric acid 33%
Catalog # FFCHFF14625000
Supplier:  FF CHEMICALS
CAS Number:  
Hydrochloric acid 33%
Catalog # FFCHFF14625000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324