Hydrochloric acid 2 mol/l (2 N)
Catalog # FFCHFF2301000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324