Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid 1 mol/l (1 N)
Hydrochloric acid 1 mol/l (1 N)
Catalog # FFCHFF2261000
Leverantör:  FF CHEMICALS
CAS-nummer:  
Hydrochloric acid 1 mol/l (1 N)
Catalog # FFCHFF2261000
Leverantör:  FF CHEMICALS
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifikationer

Om denna produkt

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
FN:1789
 :GHS05
Förvaringstemperatur:Ambient
CAS-nummer:7647-01-0
Formel:HCl
MDL-nummer:MFCD00011324