Order Entry
Sweden
ContactUsLinkComponent
Polypropylene glycol, M.W. 420
Polypropylene glycol, M.W. 420
Catalog # ROTH9148.2
Supplier:  Roth Carl
CAS Number:  
Polypropylene glycol, M.W. 420
Catalog # ROTH9148.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :1025
Storage Temperature:Ambient
EINECS:500-039-8
CAS Number:25322-69-4
Formula:H(OCH₂CH₂CH₂)nOH
MDL Number:MFCD00084446