0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Sodium thiosulphate
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF1469010
:
FF CHEMICALS
:
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF1469010
:
FF CHEMICALS
:
Pk:
10 L
Seq:
5214
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499