Order Entry
Sweden
ContactUsLinkComponent
Sodium hypochlorite (10% Cl₂) in aqueous solution
Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # FFCHVM0019030
Supplier:  FF CHEMICALS
CAS Number:  
Sodium hypochlorite (10% Cl₂) in aqueous solution
Catalog # FFCHVM0019030
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    30 L
  • Seq:
    5352

Specifications

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.22…1.25 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-668-3
UN:1791
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120