Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodine
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Iodine 0.05 mol/l (0.1 N) in aqueous solution
FFCHFF0660050
:
FF CHEMICALS
:
Iodine 0.05 mol/l (0.1 N) in aqueous solution
FFCHFF0660050
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
50 mL
Seq:
2370
:
:
253.81 g/mol
:
Ambient
:
231-442-4
:
3264
:
8,II
:
7553-56-2
:
I₂
:
MFCD00011355