Organic reference standard, Ethanol absolute 2,000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA13223000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:64-17-5
- Description:Ethanol absolute 2,000 µg/ml in methanol
- Formula:C₂H₆O
- Description:Ethanol absolute