Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium thiocyanate
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF61940500
:
FF CHEMICALS
:
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
FFCHFF61940500
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
76.12 g/mol
:
Ambient
:
217-175-6
:
1762-95-4
:
NH₄SCN
:
MFCD00011428