Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isopropyl-m-cresol
4-Isopropyl-m-cresol
4-Isopropyl-m-cresol
4-Isopropyl-m-cresol
Catalog # EHERC14463650
Supplier:
EHRENSTORFER
CAS Number:
Expand image
4-Isopropyl-m-cresol
Catalog # EHERC14463650
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pk:
0,1 g
Seq:
3020
Specifications
About this item
Product Details
:
:
150.22
EINECS:
221-761-7
CAS Number:
3228-02-2
Formula:
C₁₀H₁₄O
MDL Number:
MFCD00010704
Product Details