0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nonyn-1-ol
2-Nonyn-1-ol 96%
2-Nonyn-1-ol 96%
:
Thermo Fisher Scientific
2-Nonyn-1-ol 96%
:
Thermo Fisher Scientific
:
5921-73-3
:
:
C₉H₁₆O
:
MFCD00041566
:
140.23 g/mol
:
109…111 °C (11 mmHg)
:
0.873
:
Ambient
:
227-646-8
msds_ALFAL06380
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years