
4-Nitrophthalonitrile 99%, Sigma-Aldrich®
Catalog # 330590-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance (Colour):White to Tan
- Appearance (Form):Conforms to Requirements
Powder, Crystals, Crystalline Powder and/or Chunks - Proton NMR spectra:Conforms to Structure
- Carbon:54.7 - 56.3%
- Nitrogen:23.9 - 24.6%
- Purity (HPLC):≥98.5%
Specification Test Results
About this item
:
:173.13 g/mol
Melting Pt:144 °C
EINECS:250-748-9
:GHS07
Storage Temperature:Room Temperature
CAS Number:31643-49-9
Formula:C₈H₃N₃O₂
MDL Number:MFCD00040301