Kundt's tubes
Catalog # 765-1179
Supplier: PHYWE
Specifications
- Description:Kundts Tube
- Pk:1
- Ø×L:38x640 mm
Specifications
About this item
Glass tube used to demonstrate standing waves and acoustical forces. When used in conjunction with a sound source (single frequency), plunger to alter the effective length of the tube and cork dust or talcum powder, the tube can be made to resonate and the powder will aggregate at nodes in the tube. Calculation of the wavelength and speed of sound can be calculated from the measurement of these nodes.
Ordering information: Only glass tube supplied.