Interference tube according to quincke
Catalog # 765-1177
Supplier: PHYWE
Specifications
- Description:Quincke's pipes
- Pk:1
Specifications
About this item
Quincke interference tube for determining wavelengths and frequencies by interfering with sound waves in air. Consists of two U-shaped metal tubes slidable into each other with a measuring scale. Inputs and outputs with knobs. Three brackets for mounting on a stand. Comes with listening hose.
- Frequency range: 2-5 kHz
- Dimensions: 930 × 250 mm