Metal cylinders, same volume
Katalog # 765-0113
Leverantör: CORNELSEN EXPERIMENTA
Specifikationer
- Description:Metal cylinders, same volume
- Pk:1
Specifikationer
Om denna produkt
Rate of 6 pcs. metal cylinders of different materials. Al, Cu, Fe, Ms, Pb, and Sn. The cylinders have the same volume but different mass.