Order Entry
Sweden
ContactUsLinkComponent
Melting gutter
Katalog # 765-0206
undefined
Melting gutter
Katalog # 765-0206
Specifications
  • Description:
    Melting gutter
  • Pk:
    1
Specifications
About this item
Circular gutter made of aluminum sheet with insulating handle. Fits U-core 765-1077 and was used, among other things, when demonstrating the principle of induction furnaces.
  • Gutter: Diam, outer × inner: 95 × 70 mm
Handle: Length × Diam: 100 × 10 mm