Order Entry
Sweden
ContactUsLinkComponent
Electroscope
Electroscope
  765-0181
undefined
Electroscope
  765-0181
  • Description:
    Electroscope type Braun
  • Pk:
    1
For electrostatic experiments. Consists of insulated aluminum holder with pointed bearing, removable capacitor plate, metal ring with rod and 4 mm earthing contact.
  • Dimension: metal ring diam. 200 mm, rod diam. 10 mm