Order Entry
Sweden
ContactUsLinkComponent
Neon lamps
Neon lamps
Catalog # 765-1203
Leverantör:  PHYWE
undefined
Neon lamps
Catalog # 765-1203
Leverantör:  PHYWE
Specifikationer
  • Description:
    Neon tube
  • Pk:
    1
Specifikationer
Om denna produkt
Tubular glow lamp of glass. With conical contact hats at the ends. Used, inter alia, to determine the polarity of charge, because light only occurs around the negative electrode. Ignition voltage approximately 80 V, switching voltage approximately 65 V. For example, connects with crocodile clips.
Length: 50 mm
Ø: 10 mm