Needle holder with pin
Catalog # 765-0191
Supplier: CORNELSEN EXPERIMENTA
Specifications
- Description:Needle holder with pin
- Pk:1
Specifications
About this item
For attaching an electric spinner to the ball generator's ball. Equipped with 4 mm banana plug.