Order Entry
Sweden
ContactUsLinkComponent
Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™
Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™
Catalog # 38050-1EA
CAS Number:  
undefined
Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™
Catalog # 38050-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
ADR:8,III
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152