Order Entry
Sweden
ContactUsLinkComponent
 
Sodium periodate ≥99.8% ACS, Fluka™
 
Sodium periodate ≥99.8% ACS, Fluka™
 

 

  • Assay (excluding dried substances):
    99.8 - 100.3 %
  • Manganese (Mn):
    Max. 3.0 ppm
  • Other halogens (as Cl):
    Max. 0.02 %