Naphthalene, Supelco®
Katalog # 01422-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:128.17 g/mol
Merck Index:12,06457
Boiling Pt:218 °C (1013 hPa)
Melting Pt:79…82 °C
Density:1.145 g/cm³ (20 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:202-049-5
UN:1334
ADR:4.1,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
:GHS09
CAS Number:91-20-3
Formula:C₁₀H₈
MDL Number:MFCD00001742