Benzothiazole
Glass bottle for solids, 1 g
ROTH5689.1
Marketsource
: Benzo[d]thiazole
- :95-16-9
- :C₇H₅NS
- :MFCD00005775
- :135.19 g/mol
- :13,01108
- :231 °C (1013 hPa)
- :1.246 g/cm³ (20 °C)
- :202-396-2
- :2810
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:1 g