Order Entry
Sweden
ContactUsLinkComponent
1,3-Diethyl-2-thiourea
1,3-Diethyl-2-thiourea
Catalog # MOLE34783871-1KG
Supplier:  Molekula
CAS Number:  
1,3-Diethyl-2-thiourea
Catalog # MOLE34783871-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :132.23 g/mol
Boiling Pt:169 °C (1013 hPa)
Melting Pt:75…78 °C
Density:1,118 g/cm³ (20 °C)
Flash Pt:186 °C
EINECS:203-308-5
CAS Number:105-55-5
Formula:C₅H₁₂N₂S
MDL Number:MFCD00004925