Order Entry
Sweden
ContactUsLinkComponent
α-Bromo-p-toluonitrile
α-Bromo-p-toluonitrile
Katalog # MOLE42305814-5G
Supplier:  Molekula
CAS Number:  
α-Bromo-p-toluonitrile
Katalog # MOLE42305814-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :196.05 g/mol
Boiling Pt:135…137 °C (0.6 mmHg)
Melting Pt:115 °C
EINECS:241-246-0
UN:2923
ADR:8,II
CAS Number:17201-43-3
Formula:C₈H₆BrN
MDL Number:MFCD00001829