
Triethyl orthoformate
Catalog # MOLEM29816389
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:148.2 g/mol
Merck Index:12,07012
Boiling Pt:146 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.8964 g/cm³ (20 °C)
Flash Pt:35 °C
Storage Temperature:Ambient
EINECS:204-550-4
UN:2524
ADR:3,III
:GHS02
CAS Number:122-51-0
Formula:C₇H₁₆O₃
MDL Number:MFCD00009230