Order Entry
Sweden
ContactUsLinkComponent
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Health
Irritant
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    102.13 g/mol
  • Boiling Point:
    178 °C (1013 hPa)
  • Vapor Pressure:
    1 hPa (25 °C)
  • Flash Point:
    75 °C
  • Explosion Limit:
    1.5 - 9.7 %(V)
  • Density:
    1.05 g/cm3 (20 °C)
  • Ignition Point:
    282 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number