


(±)-2-Methyltetrahydrofuran stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:96-47-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00005367
:86.13 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:–136 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:–12 °C (closed cup)
EINECS:202-507-4
UN:2536
ADR:3,II
:M-Clarity™ quality level = MQ200
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:150 g/l (25 °C)
- Melting Point:-136 °C
- Molar Mass:86.13 g/mol
- Boiling Point:78 - 80 °C (1013 hPa)
- Vapor Pressure:136 hPa (20 °C)
- Flash Point:-11 °C
- Refractive Index:1.4056 (20 °C, 589 nm)
- Explosion Limit:1.2 - 5.7 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:270 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid