
Sodium hydroxide solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:119…135 °C (1013 hPa)
:9…10 °C
:1,53 g/cm³ (20 °C)
:215-185-5
:8,
:Filtered through a 0.2 µm filter.
:1824
:GHS05
:Ambient
- Assay (acidimetric,NaOH):32.0 - 33.0 %
- Carbonate (as Na₂CO₃):≤ 1.0 %
- Chloride (Cl):≤ 5 ppm
- Sulfate (SO₄):≤ 30 ppm
- Heavy metals (as Pb):≤ 5.0 ppm
- Al (Aluminium):≤ 10 ppm
- Fe (Iron):≤ 3 ppm
- Ni (Nickel) *:≤ 2.00 ppm
- Residual solvents (ICH Q3C):...excluded by production process
- Endotoxins:≤ 2.0 I.U./ml