Tricine, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:5704-04-1
:
Formula:(HOCH₂)₃CNHCH₂CO₂H
MDL Number:MFCD00004277
:179.17 g/mol
Merck Index:13,09724
Melting Pt:187 °C
Density:1.523 g/cm³ (19.85 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:227-193-6