Order Entry
Sweden
ContactUsLinkComponent
 
Sodium perborate tetrahydrate, pure, Supelco®
  1.06560.9050
 :  Merck
 :  
Sodium perborate tetrahydrate, pure, Supelco®
  1.06560.9050
 :  Merck
 :  

 

  • Pack type:
    Fibre carton
  • Pk:
    50 kg
  • Seq:
    4418

 

 

  • Solubility:
    23 g/l (20 °C)
  • Melting Point:
    60 °C (decomposition)
  • Molar Mass:
    153.88 g/mol
  • Bulk Density:
    800 kg/m3
  • Density:
    1.73 g/cm3
  • pH:
    10 (15 g/l, H2O, 20 °C)

 

 

 :
 :171.87 g/mol
 :60 °C
 :1.73 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
 :239-172-9
 :GHS05
 :GHS08
 :GHS07
 :Ambient
 :10486-00-7
 :NaH₂BO₄·4H₂O
 :MFCD00149231