Kromtrioxid
Catalog # LBSE790858
Supplier: LABSERVICE
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:500 mL
- Seq:2725
Specifications
About this item
:
:99.99 g/mol
Merck Index:13,02256
Melting Pt:195 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:215-607-8
UN:1463
ADR:5.1,II
REACH:01-2119458868-17
:GHS03
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:1333-82-0
Formula:CrO₃
MDL Number:MFCD00010952