

(±)-2-Methyltetrahydrofuran ≥98.0% (by GC) stabilised
: TCI
:96-47-9
:
:C₅H₁₀O
:MFCD00005367
:86.13 g/mol
:78°C
:–136 °C
:0,86 g/cm³ (20 °C)
:–11°C (closed cup)
:2536
:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:202-507-4
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %