Order Entry
Sweden
ContactUsLinkComponent
Potassium iodide 3 mol/l (3 N) in aqueous solution
Potassium iodide 3 mol/l (3 N) in aqueous solution
Catalog # FFCHFF60509010
Supplier:  FF CHEMICALS
CAS Number:  
Potassium iodide 3 mol/l (3 N) in aqueous solution
Catalog # FFCHFF60509010
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405