Order Entry
Sweden
ContactUsLinkComponent
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF20515000
Supplier:  FF CHEMICALS
CAS Number:  
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF20515000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672