Potassium iodide 1 mol/l (1 N) in aqueous solution
Catalog # FFCHFF0739010
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405