Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Thenoic acid
3-Thenoic acid
3-Thenoic acid
3-Thenoic acid
Supplier:
Thermo Scientific
Certificates
Expand image
3-Thenoic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
88-13-1
:
Formula:
C₅H₄O₂S
MDL Number:
MFCD00005467
:
128.15 g/mol
Merck Index:
12,09415
Melting Pt:
137 °C
EINECS:
201-802-5