Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Diiodo-p-xylene
2,5-Diiodo-p-xylene
2,5-Diiodo-p-xylene
2,5-Diiodo-p-xylene
Supplier:
Thermo Scientific
Certificates
Expand image
2,5-Diiodo-p-xylene
Supplier:
Thermo Scientific
About this item
CAS Number:
1124-08-9
:
Formula:
C₈H₈I₂
MDL Number:
MFCD00829126
:
357.96 g/mol