Order Entry
Sweden
ContactUsLinkComponent
 
Bovine thrombin (from plasma)
  605157-1
 :  
Bovine thrombin (from plasma)
  605157-1
 :  

 

  • Pk:
    1 KU
  • CAS No.:
    9002-04-4
  • Source:
    Plasma
  • Species:
    Bovine
  • Form:
    Lyophilised
  • Enzyme name:
    Thrombin
  • Enzyme activity:
    ≥50 NIH units/mg
  • Formulation:
    Lyophilised from 50 mM Sodium Citrate, 0,2 M NaCl, 0,1% PEG-8000, pH 6,5