Trimethylsilyl cyanide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7677-24-9
:
Formula:C₄H₉NSi
MDL Number:MFCD00001765
:99.21 g/mol
Boiling Pt:118…119 °C
Melting Pt:11…12 °C
Density:0.783
Flash Pt:1 °C (33 °F)
Storage Temperature:Ambient
EINECS:231-657-3
UN:3383
ADR:6.1,I
:GHS02
:GHS06