Acetic acid glacial ≥99.7% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Merck Index:14,55
Boiling Pt:118.1 °C
Melting Pt:16.6 °C
Density:1.049
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS05
:GHS02