3-Thenoic acid 99%
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:88-13-1
:
Formel:C₅H₄O₂S
MDL-nummer:MFCD00005467
:128.15 g/mol
Merck Index:12,09415
Smält Pt:137…139 °C
EINECS:201-802-5
:GHS07
Specifikationstestresultat
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:137 to 141 °C
- Titration with NaOH:≥98.5 %